missing translation for 'onlineSavingsMsg'
Learn More

4-[(Ethoxycarbonyl)methyl]phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number 11916101
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
859169-20-3
C16H23BO4
290.17
MFCD08704684
RPRBNRDIESZHFL-UHFFFAOYSA-N
ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenyl boronic acid pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid, pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid pinacol ester, ethyl 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenylboronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid pinacol ester, 4-2-ethoxy-2-oxoethyl phenyl boronic acid pinacol ester
46737998
ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
CCOC(=O)CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
859169-20-3
C16H23BO4
290.17
MFCD08704684
RPRBNRDIESZHFL-UHFFFAOYSA-N
ethyl 2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenyl boronic acid pinacol ester, 4-ethoxycarbonylmethylphenyl boronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid, pinacol ester, 4-ethoxycarbonylmethylphenylboronic acid pinacol ester, ethyl 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 4-ethoxycarbonylmethylphenylboronic acid, pinacol ester, 4-ethoxycarbonylmethyl phenylboronic acid pinacol ester, 4-2-ethoxy-2-oxoethyl phenyl boronic acid pinacol ester
46737998
ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
CCOC(=O)CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 41°C to 43°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 290.16
Percent Purity 97%
Chemical Name or Material 4-[(Ethoxycarbonyl)methyl]phenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 4-¢(Ethoxycarbonyl)methyl!phenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'