missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog Number p-7080693
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
6214-44-4
C9H12O2
152.19
MFCD00004655
UKFLLQIRBABMKF-UHFFFAOYSA-N
4-ethoxybenzyl alcohol, 4-ethoxyphenyl methanol, p-ethoxybenzyl alcohol, 4-ethoxybenzylalcohol, benzenemethanol, 4-ethoxy, 4-ethoxy-benzyl alcohol, benzenemethanol,4-ethoxy, 4-ethoxyphenyl methanol #, 4-ethoxyphenyl methan-1-ol, 4-ethyloxy phenyl methanol
80345
(4-ethoxyphenyl)methanol
CCOC1=CC=C(CO)C=C1
This item is not returnable. View return policy
6214-44-4
C9H12O2
152.19
MFCD00004655
UKFLLQIRBABMKF-UHFFFAOYSA-N
4-ethoxybenzyl alcohol, 4-ethoxyphenyl methanol, p-ethoxybenzyl alcohol, 4-ethoxybenzylalcohol, benzenemethanol, 4-ethoxy, 4-ethoxy-benzyl alcohol, benzenemethanol,4-ethoxy, 4-ethoxyphenyl methanol #, 4-ethoxyphenyl methan-1-ol, 4-ethyloxy phenyl methanol
80345
(4-ethoxyphenyl)methanol
CCOC1=CC=C(CO)C=C1

It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 32°C to 34°C
Boiling Point 272°C to 274°C
Flash Point >110°C (230°F)
Refractive Index 1.532
Quantity 10 g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-Ethoxybenzyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'