missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7082056
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
619-86-3
C9H10O3
166.176
MFCD00002545
SHSGDXCJYVZFTP-UHFFFAOYSA-N
p-ethoxybenzoic acid, benzoic acid, 4-ethoxy, benzoic acid, p-ethoxy, 4-ethoxy-benzoic acid, unii-tzr23xeq7w, tzr23xeq7w, 4-ethoxybenzoicacid, 4ethoxybenzoic acid, pubchem2686, para-ethoxybenzoic acid
12093
4-ethoxybenzoic acid
CCOC1=CC=C(C=C1)C(=O)O
This item is not returnable. View return policy
619-86-3
C9H10O3
166.176
MFCD00002545
SHSGDXCJYVZFTP-UHFFFAOYSA-N
p-ethoxybenzoic acid, benzoic acid, 4-ethoxy, benzoic acid, p-ethoxy, 4-ethoxy-benzoic acid, unii-tzr23xeq7w, tzr23xeq7w, 4-ethoxybenzoicacid, 4ethoxybenzoic acid, pubchem2686, para-ethoxybenzoic acid
12093
4-ethoxybenzoic acid
CCOC1=CC=C(C=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 197°C to 199°C
Quantity 25 g
Beilstein 2207349
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 4-Ethoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'