missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzhydrazide, 98+%, Thermo Scientific Chemicals

Catalog Number 15438839
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 58586-81-5
Melting Point 124°C to 127°C
Molecular Formula C9H12N2O2
MDL Number MFCD00017074
Quantity 5 g
Beilstein 2642900
Synonym 4-ethoxybenzhydrazide, p-ethoxy benzhydrazide, 4-ethoxybenzenecarbohydrazide, acmc-20amjf, 4etophcon2, #, 4-ethoxybenzoic acid, hydrazide, benzoic acid,4-ethoxy-, hydrazide
InChI Key STTILZBQLOSDNQ-UHFFFAOYSA-N
SMILES CCOC1=CC=C(C=C1)C(=O)NN
IUPAC Name 4-ethoxybenzohydrazide
Molecular Weight (g/mol) 180.207
PubChem CID 143561
Formula Weight 180.21
Percent Purity ≥98%
Chemical Name or Material 4-Ethoxybenzhydrazide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'