missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog Number p-126146
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10031-82-0
C9H10O2
150.18
MFCD00003388
JRHHJNMASOIRDS-UHFFFAOYSA-N
benzaldehyde, 4-ethoxy, p-ethoxybenzaldehyde, ethoxybenzaldehyde, benzaldehyde, p-ethoxy, homoanisaldehyde, 4-ethyloxy benzaldehyde, unii-gow1h0f49a, fema no. 2413, 4-ethoxy-benzaldehyde, ethoxybenzaldehyde, p
24834
4-ethoxybenzaldehyde
CCOC1=CC=C(C=O)C=C1
This item is not returnable. View return policy
10031-82-0
C9H10O2
150.18
MFCD00003388
JRHHJNMASOIRDS-UHFFFAOYSA-N
benzaldehyde, 4-ethoxy, p-ethoxybenzaldehyde, ethoxybenzaldehyde, benzaldehyde, p-ethoxy, homoanisaldehyde, 4-ethyloxy benzaldehyde, unii-gow1h0f49a, fema no. 2413, 4-ethoxy-benzaldehyde, ethoxybenzaldehyde, p
24834
4-ethoxybenzaldehyde
CCOC1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 13°C to 14°C
Color Yellow to Brown
Density 1.0800g/mL
Boiling Point 255°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5OC6H4CHO
Refractive Index 1.5574 to 1.5594
Quantity 5 g
Beilstein 08,73
Specific Gravity 1.08
Formula Weight 150.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Ethoxybenzaldehyde, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'