missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethoxy-3-nitropyridine hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Ethoxy-3-nitropyridine hydrochloride acts as an intermediate used in organic synthesis. It is also used in the preparation of 3,4-diaminopyridine.
Solubility
Soluble in water.
Notes
Hygroscopic and moisture sensitive.Incompatible with oxidizing agent and strong reducing agents.
4-Ethoxy-3-nitropyridine hydrochloride acts as an intermediate used in organic synthesis. It is also used in the preparation of 3,4-diaminopyridine.
Solubility
Soluble in water.
Notes
Hygroscopic and moisture sensitive.Incompatible with oxidizing agent and strong reducing agents.
Specifications
Melting Point | ∼270°C |
Quantity | 5 g |
Beilstein | 3712456 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water. |
Formula Weight | 204.61 |
Percent Purity | 97% |
Chemical Name or Material | 4-Ethoxy-3-nitropyridine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'