missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-methoxybenzaldehyde, 95%, Thermo Scientific™

Catalog Number p-126136
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
120-25-2
MFCD00016614
BERFDQAMXIBOHM-UHFFFAOYSA-N
4-ethoxy-3-anisaldehyde, 4-ethoxy-m-anisaldehyde, benzaldehyde, 4-ethoxy-3-methoxy, ethyl-vanillin, asischem n42243, 4-ethoxy-3-methoxy-benzaldehyde, ethylvanillin?, 4-ethoxyvanillin, vanillin ethyl ether, pubchem2648
67116
4-ethoxy-3-methoxybenzaldehyde
CCOC1=C(C=C(C=C1)C=O)OC
This item is not returnable. View return policy
120-25-2
MFCD00016614
BERFDQAMXIBOHM-UHFFFAOYSA-N
4-ethoxy-3-anisaldehyde, 4-ethoxy-m-anisaldehyde, benzaldehyde, 4-ethoxy-3-methoxy, ethyl-vanillin, asischem n42243, 4-ethoxy-3-methoxy-benzaldehyde, ethylvanillin?, 4-ethoxyvanillin, vanillin ethyl ether, pubchem2648
67116
4-ethoxy-3-methoxybenzaldehyde
CCOC1=C(C=C(C=C1)C=O)OC
CAS Min % 94.0
CAS Max % 100.0
Melting Point 60.0°C to 65.0°C
Color Yellow
Boiling Point 168.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C10H12O3
Linear Formula C2H5OC6H3(OCH3)CHO
Quantity 5 g
Molecular Weight (g/mol) 180.203
Formula Weight 180.2
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Ethoxy-3-methoxybenzaldehyde, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'