missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethoxy-3-methoxybenzaldehyde, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 60.0°C to 65.0°C |
Color | Yellow |
Boiling Point | 168.0°C |
Flash Point | 110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Molecular Formula | C10H12O3 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'