missing translation for 'onlineSavingsMsg'
Learn More

4-Epianhydrotetracycline hydrochloride, 'can be used as a secondary standard', ACROS Organics™

£112.00 - £390.00

Chemical Identifiers

CAS 4465-65-0
Molecular Formula C22H22N2O7·HCl
Molecular Weight (g/mol) 462.89
MDL Number MFCD00151455
InChI Key SPFAOPCHYIJPHJ-MOMXNFOMSA-N
Synonym 4-epianhydrotetracycline hydrochloride, unii-06y7hm8dba, 06y7hm8dba, 4-epi-anhydrotetracycline hydrochloride, 4-epi-ahydrochloride, 4-epianhydrotetracycline hcl, epianhydrotetracycline hydrochloride, 4-epianhydrotetracycline hydrochloride, vetranal tm , analytical standard, 4-epianhydrotetracycline hydrochloride, european pharmacopoeia ep reference standard, 4-epianhydrotetracycline hydrochloride, united states pharmacopeia usp reference standard
PubChem CID 54682545
IUPAC Name (4R,4aS,12aR)-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
SMILES CC1=C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10073170
SDS View Documents
Acros Organics
233111000
100mg Glass bottle
£112.00
100mg
View alternative products
Save Up to 
»
Estimated Shipment: 11-10-2021
Please to purchase this item. Need a web account? Register with us today!
10751891
SDS View Documents
Acros Organics
233115000
500mg Glass bottle
£390.00
500mg
View alternative products
Save Up to 
»
Estimated Shipment: 11-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

4465-65-0
462.89
SPFAOPCHYIJPHJ-MOMXNFOMSA-N
54682545
CC1=C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C.Cl
C22H22N2O7·HCl
MFCD00151455
4-epianhydrotetracycline hydrochloride, unii-06y7hm8dba, 06y7hm8dba, 4-epi-anhydrotetracycline hydrochloride, 4-epi-ahydrochloride, 4-epianhydrotetracycline hcl, epianhydrotetracycline hydrochloride, 4-epianhydrotetracycline hydrochloride, vetranal tm , analytical standard, 4-epianhydrotetracycline hydrochloride, european pharmacopoeia ep reference standard, 4-epianhydrotetracycline hydrochloride, united states pharmacopeia usp reference standard
(4R,4aS,12aR)-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride

Specifications

4-Epianhydrotetracycline hydrochloride
Glass bottle
Orange
100mg
4465-65-0
100.0
C22H22N2O7·HCl
4-epianhydrotetracycline hydrochloride, unii-06y7hm8dba, 06y7hm8dba, 4-epi-anhydrotetracycline hydrochloride, 4-epi-ahydrochloride, 4-epianhydrotetracycline hcl, epianhydrotetracycline hydrochloride, 4-epianhydrotetracycline hydrochloride, vetranal tm , analytical standard, 4-epianhydrotetracycline hydrochloride, european pharmacopoeia ep reference standard, 4-epianhydrotetracycline hydrochloride, united states pharmacopeia usp reference standard
SPFAOPCHYIJPHJ-MOMXNFOMSA-N
(4R,4aS,12aR)-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
54682545
Powder
Authentic
− 279.00
220.0°C
97%
96.0
80% min. (as is,calculated)
MFCD00151455
Solubility in water: 0.9g/L. Other solubilities: 25g/L methanol
CC1=C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C.Cl
462.89
462.89
≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC)
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Epianhydrotetracycline hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications