missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Dodecanone, 99%

Brand:  Alfa Aesar™ L04192.18

Code : NEW

Additional Details : CAS Number : 6137-26-4 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11306836

  • £206.60 / 50g


EU Stock 6
Add to basket

Specifications

Chemical Identifiers

4-Dodecanone
C12H24O
MFCD00015300
n-Octyl n-propyl ketone
6137-26-4
184.32
1703138

Specifications

4-Dodecanone
6137-26-4
C12H24O
MFCD00015300
n-Octyl n-propyl ketone
50g
99%
184.32
1703138
65°C (0.25mmHg)
Videos
SDS