missing translation for 'onlineSavingsMsg'
Learn More

4-Dodecanone, 99%, Thermo Scientific™

Catalog Number p-7058287
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
6137-26-4
C12H24O
184.323
MFCD00015300
AVQSOIZWTINZLU-UHFFFAOYSA-N
4-dodecanone, octyl propyl ketone, acmc-1bh4v
80226
dodecan-4-one
CCCCCCCCC(=O)CCC
This item is not returnable. View return policy
6137-26-4
C12H24O
184.323
MFCD00015300
AVQSOIZWTINZLU-UHFFFAOYSA-N
4-dodecanone, octyl propyl ketone, acmc-1bh4v
80226
dodecan-4-one
CCCCCCCCC(=O)CCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.83
Boiling Point 65°C (0.25mmHg)
Refractive Index 1.431
Quantity 10 g
Beilstein 1703138
Formula Weight 184.32
Percent Purity 99%
Chemical Name or Material 4-Dodecanone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'