missing translation for 'onlineSavingsMsg'
Learn More

4-(Diphenylamino)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 15432289
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
4181-05-9
C19H15NO
273.34
MFCD00145131
UESSERYYFWCTBU-UHFFFAOYSA-N
4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
77846
4-(N-phenylanilino)benzaldehyde
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O
This item is not returnable. View return policy
4181-05-9
C19H15NO
273.34
MFCD00145131
UESSERYYFWCTBU-UHFFFAOYSA-N
4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
77846
4-(N-phenylanilino)benzaldehyde
C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 129°C to 133°C (literature)
Color Yellow
Odor Odorless
Quantity 1 g
Beilstein 2732795
Sensitivity Air sensitive
Solubility Information Soluble in methanol.
Formula Weight 273.33
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(Diphenylamino)benzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'