missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminopyridine, 99%, Thermo Scientific Chemicals

Catalog Number 10504081
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
1122-58-3
MFCD00006418
VHYFNPMBLIVWCW-UHFFFAOYSA-N
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
14284
N,N-dimethylpyridin-4-amine
CN(C)C1=CC=NC=C1
This item is not returnable. View return policy
1122-58-3
MFCD00006418
VHYFNPMBLIVWCW-UHFFFAOYSA-N
4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
14284
N,N-dimethylpyridin-4-amine
CN(C)C1=CC=NC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 110.0°C to 113.0°C
Boiling Point 162.0°C (50.0 mmHg)
Flash Point 124°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H10N2
Linear Formula (C5H4N)N(CH3)2
Quantity 5 g
Beilstein 22, II, 341
Fieser 03,118; 09,178; 10,155; 12,199; 14,132; 15,131; 16,142
Merck Index 15, 3433
Solubility Information Solubility in water: 76g/L (25°C). Other solubilities: soluble in ether, slightly soluble in alcohol, benzene and, chloroform
Molecular Weight (g/mol) 122.17
Formula Weight 122.17
Vapor Pressure Negligible
Percent Purity 99%
Chemical Name or Material 4-Dimethylaminopyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Dimethylaminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'