missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3778355
19.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
619-84-1
C9H11NO2
165.19
MFCD00002537
YDIYEOMDOWUDTJ-UHFFFAOYSA-N
4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid
12092
4-(dimethylamino)benzoic acid
CN(C)C1=CC=C(C=C1)C(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
619-84-1
C9H11NO2
165.19
MFCD00002537
YDIYEOMDOWUDTJ-UHFFFAOYSA-N
4-dimethylamino benzoic acid, benzoic acid, 4-dimethylamino, benzoic acid, p-dimethylamino, p-dimethylaminobenzoic acid, n,n-dimethyl-p-aminobenzoic acid, n,n-dimethyl-4-aminobenzoic acid, p-n,n-dimethylamino benzoic acid, p-dimethylamino benzoic acid, unii-d1ma908ev0, 4-n,n-dimethylamino benzoic acid
12092
4-(dimethylamino)benzoic acid
CN(C)C1=CC=C(C=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 238°C to 242°C
Color Beige-White to Gray-Green
Density 170g/mL
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula (CH3)2NC6H4CO2H
Quantity 25 g
Beilstein 14, 426
Merck Index 15, 3254
Solubility Information Solubility in water: .. Other solubilities: soluble in hcl and koh solutions,soluble in alcohol,sparingly soluble in ether,practically insoluble in acetic acid
Formula Weight 165.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Dimethylaminobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'