missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzaldehyde, 99+%, Thermo Scientific Chemicals

Catalog Number p-4510124
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
100-10-7
C9H11NO
149.19
MFCD00003381
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
100-10-7
C9H11NO
149.19
MFCD00003381
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
7479
4-(dimethylamino)benzaldehyde
CN(C)C1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70°C to 75°C
Color Green to Yellow
Boiling Point 176°C to 177°C (17.0 mmHg)
Flash Point 164°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3)2NC6H4CHO
Quantity 25 g
Beilstein 14,31
Fieser 01,273; 02,146
Merck Index 15,3252
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in acetic acid,soluble in alcohol,ether and chloroform
Formula Weight 149.19
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Dimethylaminobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'