missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminoantipyrine, 97%, Thermo Scientific Chemicals

Catalog Number 15119163
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
58-15-1
C13H17N3O
231.30
MFCD00003142
RMMXTBMQSGEXHJ-UHFFFAOYSA-N
aminophenazone, amidopyrine, aminopyrine, 4-dimethylaminoantipyrine, dipyrine, amidazophen, amidophen, aminopyrin, pyramidon, amidophenazone
6009
CHEBI:160246
4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1
This item is not returnable. View return policy
58-15-1
C13H17N3O
231.30
MFCD00003142
RMMXTBMQSGEXHJ-UHFFFAOYSA-N
aminophenazone, amidopyrine, aminopyrine, 4-dimethylaminoantipyrine, dipyrine, amidazophen, amidophen, aminopyrin, pyramidon, amidophenazone
6009
CHEBI:160246
4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 105°C to 110°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Plastic bottle
Quantity 1 kg
Beilstein 24,46
Merck Index 15,47
Solubility Information Solubility in water: 1g/18mL
Formula Weight 231.3
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 4-Dimethylaminoantipyrine
Show More Show Less
compliance-icons
Product Identifier
  • 4-Dimethylaminoantipyrine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'