missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylamino)benzeneboronic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-(Dimethylamino)phenylboronic acid is a useful reagent for Suzuki-Miyaura cross-coupling reactions, Rhodium (Rh)-catalyzed asymmetric addition reactions, Palladium (Pd)-catalyzed C-OH bond activation. This reagent is used in Preparation of Push-pull arylvinyldiazine chromophores, Helical ortho-phenylene oligomers with terminal push-pull substitution, Alginate-supported cation-Pd nanoparticle gels as catalysts for the Suzuki-Miyaura cross-coupling reaction, Organic dyes for dye-sensitized solar cells.
Solubility
Sparingly soluble in water. (∼2.5%)
Notes
Store at room temperature. Avoid strong oxidizing agents.
4-(Dimethylamino)phenylboronic acid is a useful reagent for Suzuki-Miyaura cross-coupling reactions, Rhodium (Rh)-catalyzed asymmetric addition reactions, Palladium (Pd)-catalyzed C-OH bond activation. This reagent is used in Preparation of Push-pull arylvinyldiazine chromophores, Helical ortho-phenylene oligomers with terminal push-pull substitution, Alginate-supported cation-Pd nanoparticle gels as catalysts for the Suzuki-Miyaura cross-coupling reaction, Organic dyes for dye-sensitized solar cells.
Solubility
Sparingly soluble in water. (∼2.5%)
Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications
Melting Point | 227°C |
Quantity | 1 g |
Formula Weight | 165 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 4-(Dimethylamino)benzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'