missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Dimethylamino)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H55723.06

Code : NEW

Additional Details : CAS Number : 28611-39-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15402058

  • £95.30 / 5g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Description

Description

Applications
4-(Dimethylamino)phenylboronic acid is a useful reagent for Suzuki-Miyaura cross-coupling reactions, Rhodium (Rh)-catalyzed asymmetric addition reactions, Palladium (Pd)-catalyzed C-OH bond activation. This reagent is used in Preparation of Push-pull arylvinyldiazine chromophores, Helical ortho-phenylene oligomers with terminal push-pull substitution, Alginate-supported cation-Pd nanoparticle gels as catalysts for the Suzuki-Miyaura cross-coupling reaction, Organic dyes for dye-sensitized solar cells.

Solubility
Sparingly soluble in water. (~2.5%)

Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications

Chemical Identifiers

4-(Dimethylamino)benzeneboronic acid
165
4-(Dimethylamino)phenylboronic acid
28611-39-4
MFCD01074642

Specifications

4-(Dimethylamino)benzeneboronic acid
227°C
Solid
165
4-(Dimethylamino)phenylboronic acid
28611-39-4
5g
97%
MFCD01074642
Videos
SDS