Learn More
4-(Difluoromethoxy)aniline, 98%, ACROS Organics™
Brand: Acros Organics 399910050
Code : 29
Additional Details : CAS Number : 22236-10-8 Weight : 0.05500kg Transport : UN number : 2810 Chem class : 1 Pack group : III
Quantity | 5g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
4-(Difluoromethoxy)aniline | |
97.5 | |
98% | |
CHF2OC6H4NH2 | |
MFCD00085005 |
22236-10-8 | |
100.0 | |
C7H7F2NO | |
159.13 |
Specifications
4-(Difluoromethoxy)aniline | |
>110°C | |
5g | |
97.5 | |
Glass bottle | |
98% | |
CHF2OC6H4NH2 | |
MFCD00085005 |
1.2860g/mL | |
231.0°C | |
22236-10-8 | |
100.0 | |
98% | |
C7H7F2NO | |
159.13 |