missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)benzoic acid, 99%, Thermo Scientific™

Catalog Number p-108050
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5429-28-7
MFCD00002538
LNYTUARMNSFFBE-UHFFFAOYSA-N
4-diethylamino benzoic acid, benzoic acid, 4-diethylamino, 4-n,n-diethylaminobenzoic acid, p-diethylamino benzoic acid, 4-diethylaminobenzoicacid, acmc-209lfa, 4-diethylamino benzoic, p-diethylaminobenzoic acid, 4-diethylamino benzoicacid
79480
4-(diethylamino)benzoic acid
CCN(CC)C1=CC=C(C=C1)C(=O)O
This item is not returnable. View return policy
5429-28-7
MFCD00002538
LNYTUARMNSFFBE-UHFFFAOYSA-N
4-diethylamino benzoic acid, benzoic acid, 4-diethylamino, 4-n,n-diethylaminobenzoic acid, p-diethylamino benzoic acid, 4-diethylaminobenzoicacid, acmc-209lfa, 4-diethylamino benzoic, p-diethylaminobenzoic acid, 4-diethylamino benzoicacid
79480
4-(diethylamino)benzoic acid
CCN(CC)C1=CC=C(C=C1)C(=O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 192.0°C to 195.0°C
Color Green
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C11H15NO2
Linear Formula (C2H5)2NC6H4CO2H
Quantity 1 g
Beilstein 14, 429
Molecular Weight (g/mol) 193.24
Formula Weight 193.24
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-(Diethylamino)benzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Diethylamino)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'