missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran, 96+%

Catalog Number p-107314
24.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
51325-91-8
C19H17N3O
303.35
MFCD00051341
YLYPIBBGWLKELC-RMKNXTFCSA-N
unii-9s1wxc1zgn, dcm dye, dcm1, 9s1wxc1zgn, 4-dicyanomethylene-2-methyl-6-4-dimethylaminostyryl-4h-pyran, 4-dicyanomethylene-2-methyl-6-4-dimethylamino styryl-4h-pyran, e-2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile, 2-2-4-dimethylamino phenyl ethenyl-6-methyl-4h-pyran-4-ylidene propanedinitrile, 2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile
688222
2-[2-[(E)-2-[4-(dimethylamino)phenyl]ethenyl]-6-methylpyran-4-ylidene]propanedinitrile
CC1=CC(=C(C#N)C#N)C=C(O1)C=CC2=CC=C(C=C2)N(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
51325-91-8
C19H17N3O
303.35
MFCD00051341
YLYPIBBGWLKELC-RMKNXTFCSA-N
unii-9s1wxc1zgn, dcm dye, dcm1, 9s1wxc1zgn, 4-dicyanomethylene-2-methyl-6-4-dimethylaminostyryl-4h-pyran, 4-dicyanomethylene-2-methyl-6-4-dimethylamino styryl-4h-pyran, e-2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile, 2-2-4-dimethylamino phenyl ethenyl-6-methyl-4h-pyran-4-ylidene propanedinitrile, 2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile
688222
2-[2-[(E)-2-[4-(dimethylamino)phenyl]ethenyl]-6-methylpyran-4-ylidene]propanedinitrile
CC1=CC(=C(C#N)C#N)C=C(O1)C=CC2=CC=C(C=C2)N(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 212.0°C
Color Red
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 2841
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene,toluene,chloroform
Formula Weight 303.35
Percent Purity 96+%
Grade Extra Pure
Physical Form Crystalline Needles or Powder
Chemical Name or Material 4-(Dicyanomethylene)-2-methyl-6-(p-dimethylaminostyryl)-4H-pyran, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'