missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Cyclohexylmethylsulfamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H54490.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15401256

  • £190.00 / 1g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1221823-27-3
297.176
JDFPAMLPRYKLRV-UHFFFAOYSA-N
57655465
B(C1=CC=C(C=C1)S(=O)(=O)NCC2CCCCC2)(O)O
C13H20BNO4S
MFCD20265279
4-cyclohexylmethylsulfamoyl benzeneboronic acid, 4-cyclohexylmethylsulfamoyl phenylboronic acid
[4-(cyclohexylmethylsulfamoyl)phenyl]boronic acid

Specifications

4-(Cyclohexylmethylsulfamoyl)benzeneboronic acid
1221823-27-3
MFCD20265279
JDFPAMLPRYKLRV-UHFFFAOYSA-N
[4-(cyclohexylmethylsulfamoyl)phenyl]boronic acid
57655465
97%
1g
C13H20BNO4S
4-cyclohexylmethylsulfamoyl benzeneboronic acid, 4-cyclohexylmethylsulfamoyl phenylboronic acid
B(C1=CC=C(C=C1)S(=O)(=O)NCC2CCCCC2)(O)O
297.176
297.18
Videos
SDS Documents
Certificates