missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyclohexyliminomethyl)benzeneboronic acid pinacol ester, Thermo Scientific™

Catalog Number p-7033926
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1218790-50-1
C19H28BNO2
313.248
MFCD11113042
BAVKTLABAAKQAH-UHFFFAOYSA-N
4-cyclohexyliminomethyl benzeneboronic acid pinacol ester, 1e-n-cyclohexyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, amtb116, 4-cyclohexyl iminomethylphenylboronic acid pinacol ester, 4-cyclohexyl iminomethyl phenyl boronic acid pinacol ester, e-n-cyclohexyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, 4-cyclohexyliminomethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylidene cyclohexanamine, n-cyclohexyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine
46739736
N-cyclohexyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C=NC3CCCCC3
This item is not returnable. View return policy
1218790-50-1
C19H28BNO2
313.248
MFCD11113042
BAVKTLABAAKQAH-UHFFFAOYSA-N
4-cyclohexyliminomethyl benzeneboronic acid pinacol ester, 1e-n-cyclohexyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, amtb116, 4-cyclohexyl iminomethylphenylboronic acid pinacol ester, 4-cyclohexyl iminomethyl phenyl boronic acid pinacol ester, e-n-cyclohexyl-1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine, 4-cyclohexyliminomethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylidene cyclohexanamine, n-cyclohexyl-1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanimine
46739736
N-cyclohexyl-1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanimine
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C=NC3CCCCC3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 313.24
Chemical Name or Material 4-(Cyclohexyliminomethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'