missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylhydrazine hydrochloride, 97+%, ACROS Organics™

£15.77 - £379.00

Chemical Identifiers

CAS 2863-98-1
Molecular Formula C7H7N3·ClH
Molecular Weight (g/mol) 169.61
MDL Number MFCD00673994
InChI Key UXDLLFIRCVPPQP-UHFFFAOYSA-N
Synonym 4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
PubChem CID 16212962
IUPAC Name 4-hydrazinylbenzonitrile;hydrochloride
SMILES C1=CC(=CC=C1C#N)NN.Cl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10380464
SDS View Documents
Acros Organics
392300010
1g Glass bottle
£15.77
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 4 Please to purchase this item. Need a web account? Register with us today!
10248943
SDS View Documents
Acros Organics
392300050
5g Glass bottle
£32.15
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
10627264
SDS View Documents
Acros Organics
392300250
25g Glass bottle
£104.00
25g
View alternative products
Save Up to 
»
Estimated Shipment
08-12-2020
Please to purchase this item. Need a web account? Register with us today!
13428538
View Documents
Acros Organics
392301000
100g Glass bottle
£379.00
100g
View alternative products
Save Up to 
»
Estimated Shipment
09-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

2863-98-1
169.61
UXDLLFIRCVPPQP-UHFFFAOYSA-N
16212962
C1=CC(=CC=C1C#N)NN.Cl
C7H7N3·ClH
MFCD00673994
4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
4-hydrazinylbenzonitrile;hydrochloride

Specifications

4-Cyanophenylhydrazine hydrochloride
Authentic
Brown to Orange
1g
97.0
97% min. (HPLC)
NCC6H4NHNH2·HCl
4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
UXDLLFIRCVPPQP-UHFFFAOYSA-N
4-hydrazinylbenzonitrile;hydrochloride
16212962
Powder
>110°C
Glass bottle
249.0°C
2863-98-1
100.0
C7H7N3·ClH
MFCD00673994
Solubility in water: soluble
C1=CC(=CC=C1C#N)NN.Cl
169.61
169.61
97+%
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Cyanophenylhydrazine hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS