missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylhydrazine hydrochloride, 97+%, Thermo Scientific Chemicals

Catalog Number 13428538
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2863-98-1
C7H7N3
133.15
MFCD00673994
DZUUSHCOMPROCJ-UHFFFAOYSA-N
4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
16212962
NNC1=CC=C(C=C1)C#N
This item is not returnable. View return policy
2863-98-1
C7H7N3
133.15
MFCD00673994
DZUUSHCOMPROCJ-UHFFFAOYSA-N
4-cyanophenylhydrazine hydrochloride, 4-hydrazinylbenzonitrile hydrochloride, 4-hydrazinobenzonitrile hydrochloride, 4-cyanophenylhydrazine hcl, benzonitrile, 4-hydrazino-, monohydrochloride, 4-hydrazino-benzonitrile; hydrochloride, pubchem13684, 4-cyanophenylhydrazine-hcl, acmc-1co7p, ksc205o1j
16212962
NNC1=CC=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Melting Point 249.0°C
Color Brown to Orange
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Linear Formula NCC6H4NHNH2·HCl
Quantity 100 g
Solubility Information Solubility in water: soluble
IUPAC Name 4-hydrazinylbenzonitrile;hydrochloride
Formula Weight 169.61
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material 4-Cyanophenylhydrazine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyanophenylhydrazine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'