missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyanomethyl)phenylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number p-4602042
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
138500-86-4
C14H18BNO2
243.11
MFCD02179459
URWMFRYGXSHPRV-UHFFFAOYSA-N
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl benzeneboronic acid pinacol ester, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethylphenylboronic acid pinacol ester, 4-cyanomethylphenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile
3842922
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
CC1(C)OB(OC1(C)C)C1=CC=C(CC#N)C=C1
This item is not returnable. View return policy
138500-86-4
C14H18BNO2
243.11
MFCD02179459
URWMFRYGXSHPRV-UHFFFAOYSA-N
2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethyl benzeneboronic acid pinacol ester, 4-cyanomethyl phenylboronic acid pinacol ester, 2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-cyanomethylphenylboronic acid pinacol ester, 4-cyanomethylphenylboronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-acetonitrile
3842922
2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
CC1(C)OB(OC1(C)C)C1=CC=C(CC#N)C=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Assay Percent Range 97%
Quantity 1 g
Formula Weight 243.12
Chemical Name or Material 4-(Cyanomethyl)phenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 4-(Cyanomethyl)phenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'