missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62103.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487026

  • £352.70 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester
261.1
4-Cyanomethyl-3-fluorophenylboronic acid pinacol ester

Specifications

4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester
Solid
261.1
1g
96%
4-Cyanomethyl-3-fluorophenylboronic acid pinacol ester
Videos
SDS