missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Cyanoindole, 99%

Brand:  Alfa Aesar™ H63391.06

Code : NEW

Additional Details : CAS Number : 16136-52-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15460059

  • £57.50 / 5g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Description

Description

Applications
4-Cyanoindole is used as a synthetic intermediate.

Solubility
Soluble in water.

Notes
Heat, light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.
Specifications

Chemical Identifiers

4-Cyanoindole
142.16
Indole-4-carbonitrile
16136-52-0
MFCD00152045

Specifications

4-Cyanoindole
120° to 125°C
16136-52-0
99%
MFCD00152045
Gray
5g
Solid
142.16
Indole-4-carbonitrile
Videos
SDS