missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanobenzyl alcohol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-Cyanobenzyl alcohol is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 32°C to 36°C |
Quantity | 1 g |
UN Number | UN3439 |
Solubility Information | Insoluble in water. |
Formula Weight | 133.15 |
Percent Purity | 97% |
Chemical Name or Material | 4-Cyanobenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'