missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-2701740
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
49584-26-1
C7H4ClNO2S
201.62
MFCD00052337
DBMFYTQPPBBKHI-UHFFFAOYSA-N
4-cyanobenzene-1-sulfonyl chloride, 4-cyanobenzenesulfonylchloride, p-cyanobenzenesulphonyl chloride, 4-cyano benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyano, 4-cyanophenylsulfonyl chloride, p-cyanobenzenesulfonyl chloride, 4-cyanobenzenesulphonyl chloride, 4-cyano-benzenesulfonyl chloride, 4-chlorosulfonyl benzenecarbonitrile
39497
4-cyanobenzenesulfonyl chloride
ClS(=O)(=O)C1=CC=C(C=C1)C#N
This item is not returnable. View return policy
49584-26-1
C7H4ClNO2S
201.62
MFCD00052337
DBMFYTQPPBBKHI-UHFFFAOYSA-N
4-cyanobenzene-1-sulfonyl chloride, 4-cyanobenzenesulfonylchloride, p-cyanobenzenesulphonyl chloride, 4-cyano benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyano, 4-cyanophenylsulfonyl chloride, p-cyanobenzenesulfonyl chloride, 4-cyanobenzenesulphonyl chloride, 4-cyano-benzenesulfonyl chloride, 4-chlorosulfonyl benzenecarbonitrile
39497
4-cyanobenzenesulfonyl chloride
ClS(=O)(=O)C1=CC=C(C=C1)C#N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CNC6H4SO2Cl
Quantity 1 g
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzene-1-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'