missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Cyano-L-phenylalanine, 98%

Brand:  Alfa Aesar™ H63572.03

Code : NEW

Additional Details : CAS Number : 167479-78-9 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15440329

  • £79.10 / 1g


Estimated Shipment date
from Supplier 22-02-2018
Add to basket

Description

Description

Applications
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

4-Cyano-L-phenylalanine
190.2
UN3439
167479-78-9
MFCD00270363
H-Phe(4-CN)-OH; (S)-2-Amino-3-(4-cyanophenyl)propionic acid

Specifications

4-Cyano-L-phenylalanine
Solid
190.2
UN3439
1g
167479-78-9
98%
MFCD00270363
H-Phe(4-CN)-OH; (S)-2-Amino-3-(4-cyanophenyl)propionic acid
Videos
SDS