missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.
Solubility
Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.
Solubility
Slightly soluble in water.
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications
UN Number | UN3439 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 190.2 |
Percent Purity | 98% |
Quantity | 250 mg |
Physical Form | Solid |
Chemical Name or Material | 4-Cyano-L-phenylalanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'