missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog Number 15450329
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
167479-78-9
C10H10N2O2
190.202
MFCD00270363
KWIPUXXIFQQMKN-VIFPVBQESA-N
l-4-cyanophenylalanine, 4-cyano-l-phenylalanine, s-2-amino-3-4-cyanophenyl propanoic acid, h-phe 4-cn-oh, 2s-2-amino-3-4-cyanophenyl propanoic acid, 4-cyanophenylalanine, s-2-amino-3-4-cyanophenyl propionic acid, d-4-cn-phe-oh
1501865
(2S)-2-amino-3-(4-cyanophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)C#N
This item is not returnable. View return policy
167479-78-9
C10H10N2O2
190.202
MFCD00270363
KWIPUXXIFQQMKN-VIFPVBQESA-N
l-4-cyanophenylalanine, 4-cyano-l-phenylalanine, s-2-amino-3-4-cyanophenyl propanoic acid, h-phe 4-cn-oh, 2s-2-amino-3-4-cyanophenyl propanoic acid, 4-cyanophenylalanine, s-2-amino-3-4-cyanophenyl propionic acid, d-4-cn-phe-oh
1501865
(2S)-2-amino-3-(4-cyanophenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)N)C#N

4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
UN Number UN3439
Solubility Information Slightly soluble in water.
Formula Weight 190.2
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material 4-Cyano-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'