missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4-phenylpiperidine Hydrochloride 99%, Thermo Scientific™

Catalog Number p-95177
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
51304-58-6
C12H15ClN2
222.716
MFCD00012775
CQPHZBOPSZGTJM-UHFFFAOYSA-N
4-cyano-4-phenylpiperidine hydrochloride, 4-phenylpiperidine-4-carbonitrile hydrochloride, 4-cyano-4-phenylpiperidinehydrochloride, 4-phenylpiperidine-4-carbonitrile, chloride, 4-phenylpiperidine-4-carbonitrile hcl, pubchem19077, acmc-20a3zw, 4-cyano-4-phenylpiperidine hcl, timtec-bb sbb003334, 4-phenylisonipecotonitrile hydrochloride
3084973
4-phenylpiperidine-4-carbonitrile;hydrochloride
C1CNCCC1(C#N)C2=CC=CC=C2.Cl
This item is not returnable. View return policy
51304-58-6
C12H15ClN2
222.716
MFCD00012775
CQPHZBOPSZGTJM-UHFFFAOYSA-N
4-cyano-4-phenylpiperidine hydrochloride, 4-phenylpiperidine-4-carbonitrile hydrochloride, 4-cyano-4-phenylpiperidinehydrochloride, 4-phenylpiperidine-4-carbonitrile, chloride, 4-phenylpiperidine-4-carbonitrile hcl, pubchem19077, acmc-20a3zw, 4-cyano-4-phenylpiperidine hcl, timtec-bb sbb003334, 4-phenylisonipecotonitrile hydrochloride
3084973
4-phenylpiperidine-4-carbonitrile;hydrochloride
C1CNCCC1(C#N)C2=CC=CC=C2.Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 210.0°C to 214.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Formula Weight 222.72
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyano-4-phenylpiperidine hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyano-4-phenylpiperidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'