missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Cyano-3,5-difluoropyridine, 98%

Brand:  Alfa Aesar™ H64391.03

Code : NEW

Additional Details : CAS Number : 1214377-09-9 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15481579

  • £29.89 / 1g


Estimated Shipment date
from Supplier 22-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Cyano-3,5-difluoropyridine
140.09
UN3439
1214377-09-9
MFCD13176565
3,5-Difluoropyridine-4-carbonitrile

Specifications

4-Cyano-3,5-difluoropyridine
1214377-09-9
98%
MFCD13176565
3,5-Difluoropyridine-4-carbonitrile
1g
Solid
140.09
UN3439
Off-white
Videos
SDS