missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-3,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15497876
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1003298-73-4
C13H14BF2NO2
265.07
MFCD22419275
XYMVJTWEERDWFY-UHFFFAOYSA-N
2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-3,5-difluorobenzeneboronic acid pinacol ester, 2,6-difluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzonitrile
58504590
2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=CC(F)=C(C#N)C(F)=C1
This item is not returnable. View return policy
1003298-73-4
C13H14BF2NO2
265.07
MFCD22419275
XYMVJTWEERDWFY-UHFFFAOYSA-N
2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-3,5-difluorobenzeneboronic acid pinacol ester, 2,6-difluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzonitrile
58504590
2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=CC(F)=C(C#N)C(F)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 265.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Cyano-3,5-difluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'