missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-4602043
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1035235-29-0
C13H15BFNO2
247.08
MFCD09998162
FEUINZSEHIJUBU-UHFFFAOYSA-N
3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-2-fluorophenylboronic acid, pinacol ester, 4-cyano-2-fluorophenylboronic acid pinacol ester, 4-cyano-2-fluorobenzeneboronic acid pinacol ester, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-2-fluorophenylboronicacidpinacolester, 4-cyano-2-fluorobenzeneboronic acid, pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 3-fluoro-4-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl benzonitrile, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl benzenecarbonitrile
44755210
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C#N
This item is not returnable. View return policy
1035235-29-0
C13H15BFNO2
247.08
MFCD09998162
FEUINZSEHIJUBU-UHFFFAOYSA-N
3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-2-fluorophenylboronic acid, pinacol ester, 4-cyano-2-fluorophenylboronic acid pinacol ester, 4-cyano-2-fluorobenzeneboronic acid pinacol ester, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-2-fluorophenylboronicacidpinacolester, 4-cyano-2-fluorobenzeneboronic acid, pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 3-fluoro-4-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl benzonitrile, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl benzenecarbonitrile
44755210
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 83.0°C to 84.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 247.08
Percent Purity 97%
Chemical Name or Material 4-Cyano-2-fluorophenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 4-Cyano-2-fluorophenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'