missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothioanisole, 98%, Thermo Scientific Chemicals

Catalog Number p-7081175
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
123-09-1
C7H7ClS
158.643
MFCD00013643
KIQQUVJOLVCZKG-UHFFFAOYSA-N
4-chlorothioanisole, 4-chlorophenyl methyl sulfide, p-chlorothioanisole, 1-chloro-4-methylthio benzene, p-chlorophenyl methyl sulfide, methyl 4-chlorophenyl sulfide, benzene, 1-chloro-4-methylthio, p-chlophenyl methyl sulfide, 1-chloro-4-methylsulfanyl benzene, 4-chloro thioanisole
31243
1-chloro-4-methylsulfanylbenzene
CSC1=CC=C(C=C1)Cl
This item is not returnable. View return policy
123-09-1
C7H7ClS
158.643
MFCD00013643
KIQQUVJOLVCZKG-UHFFFAOYSA-N
4-chlorothioanisole, 4-chlorophenyl methyl sulfide, p-chlorothioanisole, 1-chloro-4-methylthio benzene, p-chlorophenyl methyl sulfide, methyl 4-chlorophenyl sulfide, benzene, 1-chloro-4-methylthio, p-chlophenyl methyl sulfide, 1-chloro-4-methylsulfanyl benzene, 4-chloro thioanisole
31243
1-chloro-4-methylsulfanylbenzene
CSC1=CC=C(C=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.22
Boiling Point 223°C
Flash Point 90°C (194°F)
Odor Unpleasant
Refractive Index 1.6
Quantity 5 g
Beilstein 2041509
Formula Weight 158.65
Percent Purity 98%
Chemical Name or Material 4-Chlorothioanisole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'