missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothieno[3,2-d]pyrimidine, 97%, Maybridge

Brand:  Maybridge GK00377EA

Code : 31

Additional Details : CAS Number : 16269-66-2 Weight : 0.06000kg

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10487612

  • £56.00 / 10g


Estimated Shipment date
from Supplier 01-05-2018
Add to basket

Specifications

Chemical Identifiers

4-Chlorothieno[3, 2-d]pyrimidine
96.5
97%
C6H3ClN2S
MFCD00122144
16269-66-2
100.0
97%
170.62

Specifications

4-Chlorothieno[3, 2-d]pyrimidine
16269-66-2
100.0
97%
C6H3ClN2S
MFCD00122144
10g
96.5
Amber Glass Bottle
97%
170.62
Videos
SDS