missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothieno[2,3-d]pyrimidine, Tech., Thermo Scientific™

Catalog Number 10064662
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
14080-59-2
C6H3ClN2S
170.61
MFCD01312373
NZCRUBBNZGVREM-UHFFFAOYSA-N
736618
4-chlorothieno[2,3-d]pyrimidine
ClC1=C2C=CSC2=NC=N1
This item is not returnable. View return policy
14080-59-2
C6H3ClN2S
170.61
MFCD01312373
NZCRUBBNZGVREM-UHFFFAOYSA-N
736618
4-chlorothieno[2,3-d]pyrimidine
ClC1=C2C=CSC2=NC=N1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 4-chlorothieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine, 4-chloro, 4-chloro-thieno 2,3-d pyrimidine, 4-chlorothiopheno 2,3-d pyrimidine, pubchem14686, acmc-1bujx, ksc522g5h, 4-chloro-thieno 2,3-d-pyrimidine, thieno 2,3-d pyrimidine,4-chloro
Formula Weight 170.62
Percent Purity 97%
Grade Technical
Chemical Name or Material 4-Chlorothieno[2, 3-d]pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'