missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals

Catalog Number p-91662
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1073-67-2
C8H7Cl
138.59
MFCD00000632
KTZVZZJJVJQZHV-UHFFFAOYSA-N
4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyrene
14085
1-chloro-4-ethenylbenzene
ClC1=CC=C(C=C)C=C1
This item is not returnable. View return policy
1073-67-2
C8H7Cl
138.59
MFCD00000632
KTZVZZJJVJQZHV-UHFFFAOYSA-N
4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyrene
14085
1-chloro-4-ethenylbenzene
ClC1=CC=C(C=C)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -16°C
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 192°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC6H4Cl
Refractive Index 1.5645 to 1.5665
Quantity 10 g
Beilstein 05, II, 367
Specific Gravity 1.1
Stabilizer 0.1% tert.-butylcatechol
Formula Weight 138.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Chlorostyrene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Chlorostyrene, stabilized
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'