missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-2701724
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1679-18-1
C6H6BClO2
156.37
MFCD00039137
CAYQIZIAYYNFCS-UHFFFAOYSA-N
4-chlorophenyl boronic acid, 4-chlorobenzeneboronic acid, p-chlorophenylboronic acid, benzeneboronic acid, p-chloro, p-chlorobenzeneboronic acid, boronic acid, 4-chlorophenyl, boronic acid, p-chlorophenyl, 4-chlorophenyl boranediol, 4-chlorophenylbornic acid
74299
(4-chlorophenyl)boronic acid
OB(O)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
1679-18-1
C6H6BClO2
156.37
MFCD00039137
CAYQIZIAYYNFCS-UHFFFAOYSA-N
4-chlorophenyl boronic acid, 4-chlorobenzeneboronic acid, p-chlorophenylboronic acid, benzeneboronic acid, p-chloro, p-chlorobenzeneboronic acid, boronic acid, 4-chlorophenyl, boronic acid, p-chlorophenyl, 4-chlorophenyl boranediol, 4-chlorophenylbornic acid
74299
(4-chlorophenyl)boronic acid
OB(O)C1=CC=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 259.0°C to 262.0°C
CAS Min % 96.0
Color White to Beige
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 96% min. (HPLC)
Linear Formula ClC6H4B(OH)2
Solubility Information Solubility in water:+/- 2.5% in water
Formula Weight 156.38
Percent Purity 97%
Chemical Name or Material 4-Chlorophenylboronic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'