missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenylacetyl chloride, 96%, Thermo Scientific Chemicals

Catalog Number 10519050
137.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
25026-34-0
C8H6Cl2O
189.04
MFCD00037111
UMQUIRYNOVNYPA-UHFFFAOYSA-N
4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride
90692
2-(4-chlorophenyl)acetyl chloride
ClC(=O)CC1=CC=C(Cl)C=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
25026-34-0
C8H6Cl2O
189.04
MFCD00037111
UMQUIRYNOVNYPA-UHFFFAOYSA-N
4-chlorophenylacetyl chloride, p-chlorophenylacetyl chloride, 2-4-chlorophenyl acetyl chloride, 4-chlorobenzeneacetyl chloride, benzeneacetyl chloride, 4-chloro, 4-chlorophenylacetylchloride, acmc-1cch1, 4-chlorobenzeneacetylchloride, 4-chlorobenzenacetyl chloride, p-chlorophenyl-acetyl chloride
90692
2-(4-chlorophenyl)acetyl chloride
ClC(=O)CC1=CC=C(Cl)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
Color Brown to Yellow
CAS Max % 100.0
Density 1.2920g/mL
Boiling Point 85.0°C (1.0 mmHg)
Quantity 100 g
Flash Point >110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5495 to 1.5565
Assay Percent Range 95% min. (GC)
Specific Gravity 1.292
Linear Formula ClC6H4CH2COCl
Solubility Information Solubility in water: reacts.
Formula Weight 189.04
Percent Purity 96%
Chemical Name or Material 4-Chlorophenylacetyl chloride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'