missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl methyl sulfone, 98%, ACROS Organics™

Brand:  Acros Organics 352200050

Code : 29

Additional Details : CAS Number : 98-57-7 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10584151

  • £21.60 / 5g


Estimated Shipment
10-05-2018
Add to basket


Specifications

Chemical Identifiers

4-Chlorophenyl methyl sulfone
98-57-7
100.0
98%
190.65
98%
97.5
98%
C7H7ClO2S
MFCD00025067

Specifications

4-Chlorophenyl methyl sulfone
96.0° to 100.0°C
98%
97.5
Authentic
Crystalline Powder
98%
190.65
Off-white
5g
98-57-7
100.0
Glass bottle
98%
C7H7ClO2S
MFCD00025067
Videos
SDS