missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Chlorophenyl chloroformate

Brand:  Alfa Aesar™ 031175.18

Code : NEW

Additional Details : CAS Number : 7693-45-0 Weight : 0.05000kg Transport : UN number : 3277 Chem class : 1a Pack group : II

Quantity 50g

 View more versions of this product

Product Code. 11307428

  • £331.25 / 50g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Chlorophenyl chloroformate
191.01
UN3277
7693-45-0
MFCD00000638

Specifications

4-Chlorophenyl chloroformate
103° (218°F)
105° to 107°C (16mmHg)
7693-45-0
1.5330
191.01
UN3277
1.365g/mL
Moisture Sensitive
50g
Pungent, makes eyes water
Liquid
MFCD00000638
Videos
SDS