missing translation for 'onlineSavingsMsg'
Learn More

(4-Chlorophenyl)(4-piperidyl)methanone hydrochloride, 95%, Thermo Scientific™

Catalog Number p-2695123
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
55695-51-7
C12H15ClNO
224.71
MFCD00053028
IYGWDOXHCPQXKN-UHFFFAOYSA-O
4-chlorophenyl piperidin-4-yl methanone hydrochloride, 4-4-chlorobenzoyl piperidine hydrochloride, 4-chlorophenyl 4-piperidyl methanone hydrochloride, 4-chlorophenyl 4-piperidinyl methanone hydrochloride, 4-p-chlorobenzoyl piperidine hydrochloride, 4-4-chlorophenyl carbonyl piperidine hydrochloride, methanone, 4-chlorophenyl-4-piperidinyl-, hydrochloride, 4-chlorophenyl 4-piperidinyl methanonehydrochloride, bestipharma 579-258, 4-4-chlorobenzoyl piperidine hcl
2777114
(4-chlorophenyl)-piperidin-4-ylmethanone;hydrochloride
ClC1=CC=C(C=C1)C(=O)C1CC[NH2+]CC1
This item is not returnable. View return policy
55695-51-7
C12H15ClNO
224.71
MFCD00053028
IYGWDOXHCPQXKN-UHFFFAOYSA-O
4-chlorophenyl piperidin-4-yl methanone hydrochloride, 4-4-chlorobenzoyl piperidine hydrochloride, 4-chlorophenyl 4-piperidyl methanone hydrochloride, 4-chlorophenyl 4-piperidinyl methanone hydrochloride, 4-p-chlorobenzoyl piperidine hydrochloride, 4-4-chlorophenyl carbonyl piperidine hydrochloride, methanone, 4-chlorophenyl-4-piperidinyl-, hydrochloride, 4-chlorophenyl 4-piperidinyl methanonehydrochloride, bestipharma 579-258, 4-4-chlorobenzoyl piperidine hcl
2777114
(4-chlorophenyl)-piperidin-4-ylmethanone;hydrochloride
ClC1=CC=C(C=C1)C(=O)C1CC[NH2+]CC1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 260.16
Percent Purity 95%
Chemical Name or Material (4-Chlorophenyl)(4-piperidyl)methanone hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'