missing translation for 'onlineSavingsMsg'
Learn More

4-(chloromethyl)-2-phenyl-1,3-thiazole, 97%, Thermo Scientific™

Catalog Number p-2700777
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
4771-31-7
C10H8ClNS
209.691
MFCD00723503
SVEGSFSFMLCNFF-UHFFFAOYSA-N
4-chloromethyl-2-phenyl-1,3-thiazole, 4-chloromethyl-2-phenylthiazole, thiazole, 4-chloromethyl-2-phenyl, 2-phenyl-4-chloromethylthiazole, 4-chloromethyl-2-phenyl-thiazole, 2-phenyl-4-chloromethyl thiazole, enamine_002044, 2-phenyl-4-chloromethyl-thiazole, 2-phenyl-1,3-thiazol-4-ylmethyl chloride
236969
4-(chloromethyl)-2-phenyl-1,3-thiazole
C1=CC=C(C=C1)C2=NC(=CS2)CCl
This item is not returnable. View return policy
4771-31-7
C10H8ClNS
209.691
MFCD00723503
SVEGSFSFMLCNFF-UHFFFAOYSA-N
4-chloromethyl-2-phenyl-1,3-thiazole, 4-chloromethyl-2-phenylthiazole, thiazole, 4-chloromethyl-2-phenyl, 2-phenyl-4-chloromethylthiazole, 4-chloromethyl-2-phenyl-thiazole, 2-phenyl-4-chloromethyl thiazole, enamine_002044, 2-phenyl-4-chloromethyl-thiazole, 2-phenyl-1,3-thiazol-4-ylmethyl chloride
236969
4-(chloromethyl)-2-phenyl-1,3-thiazole
C1=CC=C(C=C1)C2=NC(=CS2)CCl
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 209.7
Percent Purity 97%
Chemical Name or Material 4-(chloromethyl)-2-phenyl-1, 3-thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'