missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)-2-methyl-1,3-thiazole hydrochloride , Tech., Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 184.09 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 4-(Chloromethyl)-2-methyl-1, 3-thiazole hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'