missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-2,5-dimethyl-1,3-oxazole, ≥97%, Maybridge

Brand:  Maybridge CC26824DA

Code : 31

Additional Details : CAS Number : 777823-76-4 Weight : 0.05100kg Transport : UN number : 3265 Chem class : 1a Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10275714

  • £175.00 / 1g


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Specifications

Chemical Identifiers

4-(Chloromethyl)-2, 5-dimethyl-1, 3-oxazole
96.5
97%
C6H8ClNO
145.59
777823-76-4
100.0
97%
Yes
MFCD03923825

Specifications

4-(Chloromethyl)-2, 5-dimethyl-1, 3-oxazole
96.5
Amber Glass Bottle
97%
C6H8ClNO
145.59
777823-76-4
100.0
97%
1g
Yes
MFCD03923825
Videos
SDS