missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-2-(3-chlorophenyl)-1,3-thiazole hydrochloride, 97+%, Thermo Scientific™

Catalog Number p-4510154
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
690632-83-8
C10H8Cl3NS
280.59
MFCD05664422
CSXLYGPSLQHDEK-UHFFFAOYSA-N
4-chloromethyl-2-3-chlorophenyl thiazole hydrochloride, 4-chloromethyl-2-3-chlorophenyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-3-chlorophenyl-1,3-thiazole-hydrogen chloride 1/1
2794670
4-(chloromethyl)-2-(3-chlorophenyl)-1,3-thiazole;hydrochloride
Cl.ClCC1=CSC(=N1)C1=CC(Cl)=CC=C1
This item is not returnable. View return policy
690632-83-8
C10H8Cl3NS
280.59
MFCD05664422
CSXLYGPSLQHDEK-UHFFFAOYSA-N
4-chloromethyl-2-3-chlorophenyl thiazole hydrochloride, 4-chloromethyl-2-3-chlorophenyl-1,3-thiazole hydrochloride, 4-chloromethyl-2-3-chlorophenyl-1,3-thiazole-hydrogen chloride 1/1
2794670
4-(chloromethyl)-2-(3-chlorophenyl)-1,3-thiazole;hydrochloride
Cl.ClCC1=CSC(=N1)C1=CC(Cl)=CC=C1
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 280.61
Percent Purity 97%
Chemical Name or Material 4-(Chloromethyl)-2-(3-chlorophenyl)-1, 3-thiazole hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'